Nutrient pad sets
Leverancier: Dr. Möller & Schmelz
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1120-1.2 1030 1090 1110 1120-0.45 1160 1180 1190 1200 1250 1260 1099 1155 1120-0.45-MN 4095 1035 1005 1091 1145 1035-H-OM 1090H-OM 1035-H 1099-H 1110-H 1191-H 1010-H 1030-H 1040-H 1070-H 1090-H 1100-H 1120-H 1140-H 1160-H 1180-H 1200-H 1220-H 1260-H 1025-H 1040-H-OM 1115-H 1113-H 5039
391-3850EA 0 EUR
391-3850 391-3853 391-3859 391-3861 391-3862 391-3866 391-3868 391-3869 391-3870 391-3875 391-3876 MOLL1099 MOLL1155 MOLL1120-0.45-MN MOLL4095 MOLL1035 MOLL1005 MOLL1091 MOLL1145 MOLL1035-H-OM MOLL1090H-OM MOLL1035-H MOLL1099-H MOLL1110-H MOLL1191-H MOLL1010-H MOLL1030-H MOLL1040-H MOLL1070-H MOLL1090-H MOLL1100-H MOLL1120-H MOLL1140-H MOLL1160-H MOLL1180-H MOLL1200-H MOLL1220-H MOLL1260-H MOLL1025-H MOLL1040-H-OM MOLL1115-H MOLL1113-H MOLL5039
Nutrient pad sets
Media voor microbiologie Geprepareerde media voor microbiologie
Nutrient pad sets (NPS) are sterile dehydrated nutrient culture media that are ready for immediate use after addition of sterile water. They are made of biologically inert cellulose cardboard that serves as a substrate for the nutrient solution. Since it does not bind the nutrients either chemically or physically, the nutrient components are completely available for the growth of microorganisms. The composition of the various nutrient solutions used for the various NPS comply with the formulations specified in the relevant standards and regulations.
  • Storage at room temperature
  • Ready for immediate use after moistening
  • Easy handling