Anti-4930567H17Rik Rabbit Polyclonal Antibody

Leverancier: Aviva Systems Biology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AVIVARP31541_P050EA 361 EUR
AVIVARP31541_P050
Anti-4930567H17Rik Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: 4930567H17Rik
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR