Anti-ERCC8 Rabbit Polyclonal Antibody

Leverancier: Aviva Systems Biology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AVIVARP31542_P050EA 361 EUR
AVIVARP31542_P050
Anti-ERCC8 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: ERCC8 (excision repair cross-complementation group 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR