Anti-ZDHHC14 Rabbit Polyclonal Antibody
Leverancier: Aviva Systems Biology
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AVIVARP42628_P050EA 361 EUR
AVIVARP42628_P050
Anti-ZDHHC14 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: ZDHHC14 (zinc finger, DHHC-type containing 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now