Anti-SYT8 Rabbit Polyclonal Antibody

Leverancier: Aviva Systems Biology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ARP66108_P050
AVIVARP66108_P050EA 376 EUR
AVIVARP66108_P050
Anti-SYT8 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: SYT8 (synaptotagmin VIII)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR