Anti-CD19 Mouse Monoclonal Antibody (Peridinin Chlorophyll/Cy5.5®) [clone: HIB19]
Leverancier: Tonbo Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
65-0199-T100 65-0199-T025
65-0199-T100EA 165 EUR
65-0199-T100 65-0199-T025
Anti-CD19 Mouse Monoclonal Antibody (Peridinin Chlorophyll/Cy5.5®) [clone: HIB19]
Antilichamen
The HIB19 antibody reacts with human CD19, a 95 kDa glycoprotein which acts as a co-receptor, along with CD21, CD81 and CD225, in support of the functional B cell receptor (BCR). This complex provides antigen-specific recognition and subsequent activation of B cells to proliferate and differentiate into antibody-secreting cells (plasma cells) or memory B cells, which are crucial for secondary antigen encounter. CD19 is a lineage-differentiation marker, as its expression is detectable at the earliest B cell stages, through development, and is finally lost upon transition to mature plasma cells.
The HIB19 antibody is widely used as a phenotypic marker for CD19 expression on B cells, as well as on dendritic cell subsets.
Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: monoclonal
Clone: HIB19
Conjugation: Peridinin Chlorophyll/Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human