Anti-TRB Armenian Hamster Monoclonal Antibody (Peridinin Chlorophyll/Cy5.5®) [clone: H57-597]
Leverancier: Tonbo Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
65-5961-U100 65-5961-U025
65-5961-U100EA 176 EUR
65-5961-U100 65-5961-U025
Anti-TRB Armenian Hamster Monoclonal Antibody (Peridinin Chlorophyll/Cy5.5®) [clone: H57-597]
Antilichamen
The H57-597 antibody is specific for the beta chain of the mouse T cell Receptor (TCR). This cell surface protein combines with a second protein chain (alpha chain) to form the alpha-beta TCR that is expressed by NK1.1+ thymocytes, NKT cells, and the majority of peripheral T cells. A small number of T cells may express an alternative heteromer of gamma/delta protein chains, known as the g/d TCR. These receptors participate in a complex with CD3, and with the co-receptors CD4 or CD8, to recognize and respond to antigens bound to MHC molecules on antigen-presenting cells. Such interactionsMore Product Information