Anti-TMPRSS4 Goat Antibody

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORIGTA305735EA 547 EUR
ORIGTA305735
Anti-TMPRSS4 Goat Antibody
Antilichamen
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: TMPRSS4 (transmembrane protease, serine 4)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR