Anti-IDH1 Mouse Monoclonal Antibody [clone: OTI2H9]
Leverancier: OriGene
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ORIGTA500610EA 616 EUR
ORIGTA500610
Anti-IDH1 Mouse Monoclonal Antibody [clone: OTI2H9]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100
Type: Primary
Antigen: IDH1 (isocitrate dehydrogenase 1 (NADP+), soluble)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human