Anti-TFR2 Mouse Monoclonal Antibody [clone: OTI2B4]
Leverancier: OriGene
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ORIGTA504563EA 622 EUR
ORIGTA504563
Anti-TFR2 Mouse Monoclonal Antibody [clone: OTI2B4]
Antilichamen
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: TFR2 (transferrin receptor 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now