Anti-LCE2B Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB1450-100 ORB1450-200
BIRBORB1450-100EA 0 EUR
BIRBORB1450-100 BIRBORB1450-200
Anti-LCE2B Rabbit Polyclonal Antibody
Antilichamen
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: LCE2B (late cornified envelope 2B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR