Anti-PSMB8 Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB1814-100 ORB1814-200
BIRBORB1814-100EA 0 EUR
BIRBORB1814-100 BIRBORB1814-200
Anti-PSMB8 Rabbit Polyclonal Antibody
Antilichamen
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: PSMB8 (proteasome (prosome, macropain) subunit, beta type, 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Specificaties

antigeen symbool PSMB8
Environmentally Preferable
Type antilichaam Primair,Proteasome (prosome, macropain) subunit, Beta type 8
Type antilichaam Primair,Proteasome (prosome, macropain) subunit, Beta type 8
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemie Yes
Immunogeen Synthetic peptide
Formulier liquid
Purification purified by Protein A chromatography
Concentratie 0,5 mg/ml
Opslagbuffer 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR