Anti-FBXW7 Rabbit polyclonal antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB1819-100 ORB1819-20 ORB1819-200 ORB1819-5
BIRBORB1819-100EA 0 EUR
BIRBORB1819-100 BIRBORB1819-20 BIRBORB1819-200 BIRBORB1819-5
Anti-FBXW7 Rabbit polyclonal antibody
Antilichamen
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: FBXW7 (F-box and WD repeat domain containing 7, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR