Anti-ACVR2B Rabbit Polyclonal Antibody
Leverancier: Biorbyt
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ORB10140-100 ORB10140-200
BIRBORB10140-100EA 0 EUR
BIRBORB10140-100 BIRBORB10140-200
Anti-ACVR2B Rabbit Polyclonal Antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: ACVR2B (activin A receptor, type IIB)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now