Anti-CD8A Mouse Monoclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB11943-100 ORB11943-500
BIRBORB11943-100EA 0 EUR
BIRBORB11943-100 BIRBORB11943-500
Anti-CD8A Mouse Monoclonal Antibody
Antilichamen
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR