Anti-IL2RA Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®)

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB12014-100 ORB12014-50
BIRBORB12014-100EA 0 EUR
BIRBORB12014-100 BIRBORB12014-50
Anti-IL2RA Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®)
Antilichamen
Type: Primary
Antigen: IL2RA (interleukin 2 receptor, alpha)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR