Anti-CD33 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Leverancier: Biorbyt
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ORB12031-100 ORB12031-50
BIRBORB12031-100EA 0 EUR
BIRBORB12031-100 BIRBORB12031-50
Anti-CD33 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antilichamen
Type: Primary
Antigen: CD33 (CD33 molecule)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human