Anti-CD45RA Mouse Monoclonal Antibody (FITC (Fluorescein))
Leverancier: Biorbyt
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ORB12068-100 ORB12068-50
BIRBORB12068-100EA 0 EUR
BIRBORB12068-100 BIRBORB12068-50
Anti-CD45RA Mouse Monoclonal Antibody (FITC (Fluorescein))
Antilichamen
Type: Primary
Antigen: CD45RA (CD45RA)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human