Anti-ICAM1 Mouse Monoclonal Antibody (FITC (Fluorescein))

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB12089-100 ORB12089-50
BIRBORB12089-100EA 0 EUR
BIRBORB12089-100 BIRBORB12089-50
Anti-ICAM1 Mouse Monoclonal Antibody (FITC (Fluorescein))
Antilichamen
Type: Primary
Antigen: ICAM1 (intercellular adhesion molecule 1)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR