Anti-FCGR1A Mouse Monoclonal Antibody (FITC (Fluorescein))

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB12116-100 ORB12116-50
BIRBORB12116-100EA 0 EUR
BIRBORB12116-100 BIRBORB12116-50
Anti-FCGR1A Mouse Monoclonal Antibody (FITC (Fluorescein))
Antilichamen
Type: Primary
Antigen: FCGR1A (Fc fragment of IgG, high affinity Ia, receptor (CD64))
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR