Anti-BSG Mouse monoclonal antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB12138-100 ORB12138-500
BIRBORB12138-100EA 0 EUR
BIRBORB12138-100 BIRBORB12138-500
Anti-BSG Mouse monoclonal antibody
Antilichamen
Type: Primary
Antigen: BSG (basigin (Ok blood group))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR