Anti-HLA-DR Mouse Monoclonal Antibody (FITC (Fluorescein))
Leverancier: Biorbyt
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ORB12151-100 ORB12151-50
BIRBORB12151-100EA 0 EUR
BIRBORB12151-100 BIRBORB12151-50
Anti-HLA-DR Mouse Monoclonal Antibody (FITC (Fluorescein))
Antilichamen
Type: Primary
Antigen: HLA-DR
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human