Anti-SLC6A4 Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB13226-100 ORB13226-20 ORB13226-200 ORB13226-5
BIRBORB13226-100EA 0 EUR
BIRBORB13226-100 BIRBORB13226-20 BIRBORB13226-200 BIRBORB13226-5
Anti-SLC6A4 Rabbit Polyclonal Antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: SLC6A4 (solute carrier family 6 (neurotransmitter transporter), member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR