Anti-ITGAM Rat Monoclonal Antibody (FITC (Fluorescein))

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB22690-1 ORB22690-500
BIRBORB22690-1EA 0 EUR
BIRBORB22690-1 BIRBORB22690-500
Anti-ITGAM Rat Monoclonal Antibody (FITC (Fluorescein))
Antilichamen
Type: Primary
Antigen: ITGAM (integrin, alpha M (complement component 3 receptor 3 subunit))
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2b (Rat)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR