Anti-HSF1 Mouse Monoclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB22744-100 ORB22744-20 ORB22744-5
BIRBORB22744-100EA 0 EUR
BIRBORB22744-100 BIRBORB22744-20 BIRBORB22744-5
Anti-HSF1 Mouse Monoclonal Antibody
Antilichamen
Type: Primary
Antigen: HSF1 (heat shock transcription factor 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR