Anti-MAT2A Mouse Monoclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB22776-100 ORB22776-20 ORB22776-5
BIRBORB22776-100EA 0 EUR
BIRBORB22776-100 BIRBORB22776-20 BIRBORB22776-5
Anti-MAT2A Mouse Monoclonal Antibody
Antilichamen
Type: Primary
Antigen: MAT2A (methionine adenosyltransferase II, alpha)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR