Anti-CALM1 Mouse Monoclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB22808-100 ORB22808-20 ORB22808-5
BIRBORB22808-100EA 0 EUR
BIRBORB22808-100 BIRBORB22808-20 BIRBORB22808-5
Anti-CALM1 Mouse Monoclonal Antibody
Antilichamen
Type: Primary
Antigen: CALM1 (calmodulin 1 (phosphorylase kinase, delta))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR