Anti-GFP Mouse monoclonal antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB22827-100 ORB22827-20 ORB22827-5
BIRBORB22827-100EA 0 EUR
BIRBORB22827-100 BIRBORB22827-20 BIRBORB22827-5
Anti-GFP Mouse monoclonal antibody
Antilichamen
Type: Primary
Antigen: GFP (green fluorescent protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR