Anti-COL1 Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB27305-0.1 ORB27305-0.5
BIRBORB27305-0.1EA 0 EUR
BIRBORB27305-0.1 BIRBORB27305-0.5
Anti-COL1 Rabbit Polyclonal Antibody
Antilichamen
ImmunofluorescenceA 1:80; Immunochemistry: 1:500; ELISA: >1:200

Type: Primary
Antigen: COL1 (collagen, type I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR