Anti-ELN Guinea pig polyclonal antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB27331-0.1 ORB27331-0.5
BIRBORB27331-0.1EA 0 EUR
BIRBORB27331-0.1 BIRBORB27331-0.5
Anti-ELN Guinea pig polyclonal antibody
Antilichamen
ImmunofluorescenceA 1:80; Immunochemistry: 1:500; ELISA: >1:200

Type: Primary
Antigen: ELN (elastin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Guinea pig
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR