Anti-SPHK1 Rabbit Polyclonal Antibody
Leverancier: Biorbyt
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ORB49084-100 ORB49084-200
BIRBORB49084-100EA 0 EUR
BIRBORB49084-100 BIRBORB49084-200
Anti-SPHK1 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: SPHK1 (sphingosine kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat