Anti-mCherry Tag Mouse Monoclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB66657-100 ORB66657-200
BIRBORB66657-100EA 0 EUR
BIRBORB66657-100 BIRBORB66657-200
Anti-mCherry Tag Mouse Monoclonal Antibody
Antilichamen
Western Blot: 1:3000-1:10000; Immunochemistry: 1:200; Immunofluorescence: 1:200

Type: Primary
Antigen: mCherry Tag
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR