Anti-RNF7 Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB100014-100 ORB100014-200
BIRBORB100014-100EA 0 EUR
BIRBORB100014-100 BIRBORB100014-200
Anti-RNF7 Rabbit Polyclonal Antibody
Antilichamen
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; IC 1:100-500; Immunofluorescence: 1:50-200

Type: Primary
Antigen: RNF7 (ring finger protein 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Specificaties

antigeen symbool RNF7
Environmentally Preferable
Type antilichaam Ring Finger Protein 7,Primair
Type antilichaam Ring Finger Protein 7,Primair
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemie Yes
Formulier liquid
Moleculair gewicht 28 kDa
Purification purified by affinity chromatography
Opslagbuffer 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR