Anti-TRAIP Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB100669-100 ORB100669-200
BIRBORB100669-100EA 0 EUR
BIRBORB100669-100 BIRBORB100669-200
Anti-TRAIP Rabbit Polyclonal Antibody
Antilichamen
Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: TRAIP (TRAF interacting protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Specificaties

antigeen symbool TRAIP
Type antilichaam Primair,TRAF interacting protein
Environmentally Preferable
Type antilichaam Primair,TRAF interacting protein
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity
Host Rabbit
Isotype IgG
ImmunoChemie Yes
ImmunoFluorescentie Yes
Formulier liquid
Moleculair gewicht 53 kDa
Purification purified by affinity chromatography
Concentratie 0,5 mg/ml
Opslagbuffer 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR