Anti-KDM4C Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB100883-100 ORB100883-200
BIRBORB100883-100EA 0 EUR
BIRBORB100883-100 BIRBORB100883-200
Anti-KDM4C Rabbit Polyclonal Antibody
Antilichamen
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunoprecipitation: 1:20-100; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: KDM4C (lysine (K)-specific demethylase 4C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR