Anti-AIM2 Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB101275-100 ORB101275-200
BIRBORB101275-100EA 0 EUR
BIRBORB101275-100 BIRBORB101275-200
Anti-AIM2 Rabbit Polyclonal Antibody
Antilichamen
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: AIM2 (absent in melanoma 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR