Anti-DAB2IP Rabbit polyclonal antibody
Leverancier: Biorbyt
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ORB101664-100 ORB101664-200
BIRBORB101664-100EA 0 EUR
BIRBORB101664-100 BIRBORB101664-200
Anti-DAB2IP Rabbit polyclonal antibody
Antilichamen
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: DAB2IP (DAB2 interacting protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now