Anti-IgG Mouse Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB101977-100 ORB101977-200
BIRBORB101977-100EA 0 EUR
BIRBORB101977-100 BIRBORB101977-200
Anti-IgG Mouse Polyclonal Antibody
Antilichamen
Western Blot: 1:1000-5000; ELISA: 1:2000-10000; Immunochemistry: 1:200-1000; IC 1:200-1000

Type: Secondary
Antigen: IgG (Guinea pig) (guinea pig IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Guinea pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR