Anti-CAMK2B Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB106287-100 ORB106287-20
BIRBORB106287-100EA 0 EUR
BIRBORB106287-100 BIRBORB106287-20
Anti-CAMK2B Rabbit Polyclonal Antibody
Antilichamen
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200; Immunofluorescence: 1:100-1:500

Type: Primary
Antigen: CAMK2B (calcium/calmodulin-dependent protein kinase II beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR