Anti-CDK1 Rabbit polyclonal antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB106340-100 ORB106340-20
BIRBORB106340-100EA 0 EUR
BIRBORB106340-100 BIRBORB106340-20
Anti-CDK1 Rabbit polyclonal antibody
Antilichamen
Western Blot: 1:500-1:2000

Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR