Anti-JUND Rabbit Polyclonal Antibody
Leverancier: Biorbyt
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ORB106389-100 ORB106389-20
BIRBORB106389-100EA 0 EUR
BIRBORB106389-100 BIRBORB106389-20
Anti-JUND Rabbit Polyclonal Antibody
Antilichamen
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200
Type: Primary
Antigen: JUND (jun D proto-oncogene)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat