Anti-PI 3 Kinase gamma Human Monoclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB122544-1 ORB122544-200
BIRBORB122544-1EA 0 EUR
BIRBORB122544-1 BIRBORB122544-200
Anti-PI 3 Kinase gamma Human Monoclonal Antibody
Antilichamen
Type: Primary
Antigen: PI 3 Kinase gamma
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Human
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR