Anti-AKR1C2 Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB125409-100 ORB125409-200
BIRBORB125409-100EA 0 EUR
BIRBORB125409-100 BIRBORB125409-200
Anti-AKR1C2 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: AKR1C2 (aldo-keto reductase family 1, member C2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Specificaties

antigeen symbool AKR1C2
Type antilichaam Primair,aldo-keto reductase family 1, member C2
Environmentally Preferable
Type antilichaam Primair,aldo-keto reductase family 1, member C2
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Human
Host Rabbit
Isotype
Western blot Yes
Purification purified by affinity chromatography
Concentratie 1 mg/ml
Opslagbuffer PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR