Anti-APOA5 Rabbit Polyclonal Antibody
Leverancier: Biorbyt
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ORB125430-100 ORB125430-200
BIRBORB125430-100EA 0 EUR
BIRBORB125430-100 BIRBORB125430-200
Anti-APOA5 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: APOA5 (apolipoprotein A-V)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human