Anti-Caspase 3 (p17) Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB126609-100 ORB126609-200
BIRBORB126609-100EA 0 EUR
BIRBORB126609-100 BIRBORB126609-200
Anti-Caspase 3 (p17) Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: Caspase 3 (p17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR