Anti-PECAM1 Rabbit Polyclonal Antibody
Leverancier: Biorbyt
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ORB126698-100 ORB126698-200
BIRBORB126698-100EA 0 EUR
BIRBORB126698-100 BIRBORB126698-200
Anti-PECAM1 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: PECAM1 (platelet/endothelial cell adhesion molecule 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now