Anti-MEKKK 1 Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB127036-100 ORB127036-200
BIRBORB127036-100EA 0 EUR
BIRBORB127036-100 BIRBORB127036-200
Anti-MEKKK 1 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: MEKKK 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Specificaties

antigeen symbool MEKKK1
Environmentally Preferable
Type antilichaam Primair
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity
Host Rabbit
Isotype
Western blot Yes
Purification purified by affinity chromatography
Concentratie 1 mg/ml
Opslagbuffer PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR