Anti-MED27 Rabbit Polyclonal Antibody
Leverancier: Biorbyt
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ORB127057-100 ORB127057-200
BIRBORB127057-100EA 0 EUR
BIRBORB127057-100 BIRBORB127057-200
Anti-MED27 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: MED27 (mediator complex subunit 27)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now