Anti-B4GALT3 Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB127740-100 ORB127740-200
BIRBORB127740-100EA 0 EUR
BIRBORB127740-100 BIRBORB127740-200
Anti-B4GALT3 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: B4GALT3 (UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR