Anti-EIF1AY Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB128011-100 ORB128011-200
BIRBORB128011-100EA 0 EUR
BIRBORB128011-100 BIRBORB128011-200
Anti-EIF1AY Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: EIF1AY (eukaryotic translation initiation factor 1A, Y-linked)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR